1-Azetidinecarboxylic acid-3-hydroxyphenylmethyl ester 96%

Lieferant: Apollo Scientific

Synonyme: Benzyl 3-hydroxyazetidine-1-carboxylate, 1-Cbz-3-Hydroxyazetidine

OR309038-25G OR309038-100G
APOSOR309038-25GEA 59.6 EUR
APOSOR309038-25G APOSOR309038-100G
1-Azetidinecarboxylic acid-3-hydroxyphenylmethyl ester 96%
1-Azetidinecarboxylic acid-3-hydroxyphenylmethyl ester
Formel: C₁₁H₁₃NO₃
Molecular Weight: 207,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09037849
CAS-Nummer: 128117-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR