3-Bromomethyl-3-methyloxetane

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Bromomethyl)-2-methyl-1,3-epoxypropane, 3-(Bromomethyl)-3-methyloxetane

OR309052-1G OR309052-5G
APOSOR309052-1GEA 26.1 EUR
APOSOR309052-1G APOSOR309052-5G
3-Bromomethyl-3-methyloxetane
3-Bromomethyl-3-methyloxetane
Formel: C₅H₉BrO
Molecular Weight: 165,04 g/mol
Siedepunkt: 162 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02684290
CAS-Nummer: 78385-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR