3-Methyl-3-oxetanmethanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Methyl-3-oxethanemethanol
OR309068-100G
OR309068-25G
OR309068-500G
APOSOR309068-100GEA
48
EUR
APOSOR309068-100G
APOSOR309068-25G
APOSOR309068-500G
3-Methyl-3-oxetanmethanol
3-Methyl-3-oxetanmethanol
Formel:
C₅H₁₀O₂ Molecular Weight: 102,13 g/mol Siedepunkt: 78…80 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00010273 CAS-Nummer: 3143-02-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...