3-(Aminomethyl)oxetan-3-ol 97%

Lieferant: Apollo Scientific

OR309100-1G OR309100-5G
APOSOR309100-1GEA 473 EUR
APOSOR309100-1G APOSOR309100-5G
3-(Aminomethyl)oxetan-3-ol 97%
3-(Aminomethyl)oxetan-3-ol
Formel: C₄H₉NO₂
Molecular Weight: 103,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20444451
CAS-Nummer: 1305208-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR