D-β-Proline 97%
Lieferant: Apollo Scientific
Synonyme:
(R)-Pyrrolidine-3-carboxylic acid, H-D-β-Pro-OH
OR309146-1G
OR309146-5G
APOSOR309146-1GEA
140
EUR
APOSOR309146-1G
APOSOR309146-5G
D-β-Proline 97%
D-beta-Proline
Formel:
C₅H₉NO₂ Molecular Weight: 115,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08272856 CAS-Nummer: 72580-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...