6-Bromo-2-chloroquinoline-3-carboxaldehyde

Lieferant: Apollo Scientific

Synonyme: 6-Bromo-2-chloroquinoline-3-carbaldehyde

OR309202-1G OR309202-5G
APOSOR309202-1GEA 173 EUR
APOSOR309202-1G APOSOR309202-5G
6-Bromo-2-chloroquinoline-3-carboxaldehyde
6-Bromo-2-chloroquinoline-3-carboxaldehyde
Formel: C₁₀H₅BrClNO
Molecular Weight: 270.51 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08706312
CAS-Nummer: 73568-35-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR