6-Chloro-2-methylquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR309207-1G OR309207-5G
APOSOR309207-1GEA 301 EUR
APOSOR309207-1G APOSOR309207-5G
6-Chloro-2-methylquinoline-3-carboxylic acid
6-Chloro-2-methylquinoline-3-carboxylic acid
Formel: C₁₁H₈ClNO₂
Molecular Weight: 221.64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06254736
CAS-Nummer: 92513-40-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR