6-Methoxy-2-methylquinoline-3-carboxylic acid
Lieferant: Apollo Scientific
OR309215-1G OR309215-5G
APOSOR309215-1GEA 317 EUR
APOSOR309215-1G APOSOR309215-5G
6-Methoxy-2-methylquinoline-3-carboxylic acid
6-Methoxy-2-methylquinoline-3-carboxylic acid
Formel: C₁₂H₁₁NO₃
Molecular Weight: 217,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00841563
CAS-Nummer: 88752-76-5