7,8-Dichlor-4-chinolinol

Lieferant: Apollo Scientific

Synonyme: 7,8-Dichloro-4-hydroxyquinoline

OR309218-1G OR309218-250MG
APOSOR309218-1GEA 284 EUR
APOSOR309218-1G APOSOR309218-250MG
7,8-Dichlor-4-chinolinol
7,8-Dichlor-4-chinolinol
Formel: C₉H₅Cl₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00272453
CAS-Nummer: 871217-91-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR