Ethyl-8-chlor-4-hydroxychinolin-3-carboxylat
Lieferant: Apollo Scientific
Synonyme:
Ethyl 8-chloro-4-hydroxyquinoline-3-carboxylate, 8-Chloro-4-hydroxyquinoline-3-carboxylic acid ethyl ester
OR309235-5G
APOSOR309235-5GEA
66.7
EUR
APOSOR309235-5G
Ethyl-8-chlor-4-hydroxychinolin-3-carboxylat
Ethyl-8-chlor-4-hydroxychinolin-3-carboxylat
Formel:
C₁₂H₁₀ClNO₃ Molecular Weight: 251,6657 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024002 CAS-Nummer: 73987-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...