6-Methoxy-2-oxo-1,2-dihydro-quinoline-3-carbaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Hydroxy-6-methoxyquinoline-3-carbaldehyde

OR309253-1G OR309253-5G
APOSOR309253-1GEA 310 EUR
APOSOR309253-1G APOSOR309253-5G
6-Methoxy-2-oxo-1,2-dihydro-quinoline-3-carbaldehyde
6-Methoxy-2-oxo-1,2-dihydro-quinoline-3-carbaldehyde
Formel: C₁₁H₉NO₃
Molecular Weight: 203.2 g/mol
Schmelzpunkt: 248…250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02227056
CAS-Nummer: 123990-78-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR