2-Chloro-6-methoxyquinoline-3-methanol

Lieferant: Apollo Scientific

OR309262-1G OR309262-100MG OR309262-250MG OR309262-500MG OR309262-2.5G
APOSOR309262-1GEA 393 EUR
APOSOR309262-1G APOSOR309262-100MG APOSOR309262-250MG APOSOR309262-500MG APOSOR309262-2.5G
2-Chloro-6-methoxyquinoline-3-methanol
2-Chloro-6-methoxyquinoline-3-methanol
Formel: C₁₁H₁₀ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02232332
CAS-Nummer: 92172-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR