3-Hydroxy-4-chinolincarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR309281-1G
OR309281-250MG
APOSOR309281-1GEA
294
EUR
APOSOR309281-1G
APOSOR309281-250MG
3-Hydroxy-4-chinolincarbonsäure
3-Hydroxy-4-chinolincarbonsäure
Formel:
C₁₀H₇NO₃ Molecular Weight: 189.17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01536959 CAS-Nummer: 118-13-8 EINECS: 204-236-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...