4-Bromo-8-methoxy-2-methylquinoline ≥97%

Lieferant: Apollo Scientific

OR309289-250MG OR309289-100MG
APOSOR309289-250MGEA 125 EUR
APOSOR309289-250MG APOSOR309289-100MG
4-Bromo-8-methoxy-2-methylquinoline ≥97%
4-Bromo-8-methoxy-2-methylquinoline
Formel: C₁₁H₁₀BrNO
Lagertemperatur: Kühlschrank
CAS-Nummer: 927800-62-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR