5-Brom-2-nitrophenol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Brom-3-hydroxy-4-nitrobenzol
OR30929-10G
OR30929-1G
OR30929-5G
APOSOR30929-10GEA
31.9
EUR
APOSOR30929-10G
APOSOR30929-1G
APOSOR30929-5G
5-Brom-2-nitrophenol 97%
5-Brom-2-nitrophenol
Formel:
C₆H₄BrNO₃ Schmelzpunkt: 40…41 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095027 CAS-Nummer: 27684-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...