4-Brom-o-anisidin 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Methoxy-4-bromanilin, 2-Amino-5-bromanisol, 4-Brom-2-methoxyanilin

OR30932-100G OR30932-25G OR30932-5G
APOSOR30932-100GEA 154 EUR
APOSOR30932-100G APOSOR30932-25G APOSOR30932-5G
4-Brom-o-anisidin 98%
4-Brom-o-anisidin
Formel: C₇H₈BrNO
Molecular Weight: 202,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01204266
CAS-Nummer: 59557-91-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR