Ethyl-4-chlor-6-methoxychinolin-3-carboxylat

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate, 4-Chloro-6-methoxyquinoline-3-carboxylic acid ethyl ester

OR309349-1G OR309349-5G
APOSOR309349-1GEA 42.2 EUR
APOSOR309349-1G APOSOR309349-5G
Ethyl-4-chlor-6-methoxychinolin-3-carboxylat
Ethyl-4-chlor-6-methoxychinolin-3-carboxylat
Formel: C₁₃H₁₂ClNO₃
Molecular Weight: 265,70 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173417
CAS-Nummer: 22931-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR