4-Brom-3-methoxyphenol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR30937-1G
OR30937-25G
OR30937-5G
APOSOR30937-1GEA
21.9
EUR
APOSOR30937-1G
APOSOR30937-25G
APOSOR30937-5G
4-Brom-3-methoxyphenol 98%
4-Brom-3-methoxyphenol
Formel:
C₇H₇BrO₂ Molecular Weight: 203.04 g/mol Schmelzpunkt: 77…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00029748 CAS-Nummer: 102127-34-4 EINECS: 634-447-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...