2,7-Dichlor-4-methylchinolin
Lieferant: Apollo Scientific
OR309382-1G
OR309382-5G
APOSOR309382-1GEA
127
EUR
APOSOR309382-1G
APOSOR309382-5G
2,7-Dichlor-4-methylchinolin
2,7-Dichloro-4-methylquinoline
Formel:
C₁₀H₇Cl₂N Lagertemperatur: Raumtemperatur |
CAS-Nummer:
59666-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...