Methyl-3-amino-2-chloroisonicotinate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-amino-2-chloroisonicotinate

OR30947-5G
APOSOR30947-5GEA 77 EUR
APOSOR30947-5G
Methyl-3-amino-2-chloroisonicotinate 95%
Methyl-3-amino-2-chloroisonicotinate
Formel: C₇H₇ClN₂O₂
Molecular Weight: 186,6000 g/mol
Schmelzpunkt: 94…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03411723
CAS-Nummer: 173435-41-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR