3-Amino-6-chloropyridine-2-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR30951-1G
OR30951-250MG
APOSOR30951-1GEA
134
EUR
APOSOR30951-1G
APOSOR30951-250MG
3-Amino-6-chloropyridine-2-carbonitrile
3-Amino-6-chloropyridine-2-carbonitrile
Formel:
C₆H₄ClN₃ Schmelzpunkt: 170…171 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07780767 CAS-Nummer: 95095-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...