3-Amino-5-hydroxy-1H-pyrazol 98%
Lieferant: Apollo Scientific
Synonyme:
3-Amino-5-hydroxy-1H-pyrazole, 3-Amino-5-hydroxypyrazole, 5-Amino-3-pyrazolol, 3-Amino-1H-pyrazol-5(4H)-one, 3-Amino-2-pyrazolin-5-one, 5-Hydroxy-3-aminopyrazole, 5-Amino-2,4-dihydro-3H-pyrazol-3-one
OR30956-100G
OR30956-25G
APOSOR30956-100GEA
212
EUR
APOSOR30956-100G
APOSOR30956-25G
3-Amino-5-hydroxy-1H-pyrazol 98%
3-Amino-5-hydroxy-1H-pyrazole
Formel:
C₃H₅N₃O Molecular Weight: 99.09 g/mol Schmelzpunkt: 214 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022384 CAS-Nummer: 6126-22-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...