3-Amino-6-methylpyridazin
Lieferant: Apollo Scientific
|
Achtung
|
OR30962-1G
OR30962-5G
APOSOR30962-1GEA
23.2
EUR
APOSOR30962-1G
APOSOR30962-5G
3-Amino-6-methylpyridazin
3-Amino-6-methylpyridazin
Formel:
C₅H₇N₃ Molecular Weight: 109.13 g/mol Schmelzpunkt: 223…227 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01569899 CAS-Nummer: 18591-82-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...