2-Butyl-1,3-benzoxazol-5-amin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Amino-2-(but-1-yl)-1,3-benzoxazole
OR30963-10G
OR30963-1G
APOSOR30963-10GEA
0
EUR
APOSOR30963-10G
APOSOR30963-1G
2-Butyl-1,3-benzoxazol-5-amin
2-Butyl-1,3-benzoxazol-5-amin
Formel:
C₁₁H₁₄N₂O Lagertemperatur: Raumtemperatur |
CAS-Nummer:
885949-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...