1-Benzyl-3-pyrrolidinamin 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Amino-1-benzylpyrrolidine, 1-Benzylpyrrolidin-3-amine

OR30970-100G OR30970-25G OR30970-5G
APOSOR30970-100GEA 754 EUR
APOSOR30970-100G APOSOR30970-25G APOSOR30970-5G
1-Benzyl-3-pyrrolidinamin 97%
1-Benzyl-3-aminopyrrolidine
Formel: C₁₁H₁₆N₂
Molecular Weight: 176,26 g/mol
Dichte: 1,024 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00059036
CAS-Nummer: 18471-40-4
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR