3-Amino-1H-indazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-1H-indazol, Indazol-3-ylamin, 1H-Indazol-3-ylamin, 1H-Indazol-3-amin, Indazol-3-amin

OR30984-1G OR30984-5G OR30984-25G
APOSOR30984-1GEA 21.9 EUR
APOSOR30984-1G APOSOR30984-5G APOSOR30984-25G
3-Amino-1H-indazol
3-Amino-1H-indazole
Formel: C₇H₇N₃
Molecular Weight: 133,15 g/mol
Schmelzpunkt: 156…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00182045
CAS-Nummer: 874-05-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR