1-Methyl-1H-pyrazole-3-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR30991-1G OR30991-5G
APOSOR30991-1GEA 21.9 EUR
APOSOR30991-1G APOSOR30991-5G
1-Methyl-1H-pyrazole-3-carbaldehyde
1-Methyl-1H-pyrazole-3-carbaldehyde
Formel: C₅H₆N₂O
Molecular Weight: 110.1100 g/mol
Siedepunkt: 88…90 °C/0.1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD03419800
CAS-Nummer: 27258-32-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR