2-Chloro-1-methyl-1H-indole-3-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR30993-1G
OR30993-250MG
APOSOR30993-1GEA
40.6
EUR
APOSOR30993-1G
APOSOR30993-250MG
2-Chloro-1-methyl-1H-indole-3-carbaldehyde
2-Chloro-1-methyl-1H-indole-3-carbaldehyde
Formel:
C₁₀H₈ClNO Molecular Weight: 193,63 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02253186 CAS-Nummer: 24279-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...