2,6-Dichloro-4-(chloromethyl)pyridine

Lieferant: Apollo Scientific
Gefahr

OR310005-1G OR310005-250MG
APOSOR310005-1GEA 87 EUR
APOSOR310005-1G APOSOR310005-250MG
2,6-Dichloro-4-(chloromethyl)pyridine
2,6-Dichloro-4-(chloromethyl)pyridine
Formel: C₆H₄Cl₃N
Molecular Weight: 196.46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03783562
CAS-Nummer: 101990-72-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR