2-Benzyloxy-3-bromopyridine

Lieferant: Apollo Scientific

OR310082-1G
APOSOR310082-1GEA 259 EUR
APOSOR310082-1G
2-Benzyloxy-3-bromopyridine
2-Benzyloxy-3-bromopyridine
Formel: C₁₂H₁₀BrNO
Molecular Weight: 264,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09037467
CAS-Nummer: 52200-49-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR