4,4-Dimethyl-1,2-oxazolidin-3-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4,4-Dimethylisoxazolidin-3-one
OR310131-1G
OR310131-100MG
OR310131-250MG
APOSOR310131-1GEA
101
EUR
APOSOR310131-1G
APOSOR310131-100MG
APOSOR310131-250MG
4,4-Dimethyl-1,2-oxazolidin-3-one
4,4-Dimethyl-1,2-oxazolidin-3-one
Formel:
C₅H₉NO₂ Molecular Weight: 115,13 g/mol Schmelzpunkt: 74…76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD21333197 CAS-Nummer: 81778-07-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...