3-Iodo-5-methylpyridin-2-ol
Lieferant: Apollo Scientific
Synonyme:
2-Hydroxy-3-iodo-5-methylpyridine
OR310163-1G
APOSOR310163-1GEA
63.8
EUR
APOSOR310163-1G
3-Iodo-5-methylpyridin-2-ol
3-Iodo-5-methylpyridin-2-ol
Formel:
C₆H₆INO Molecular Weight: 235,03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09037469 CAS-Nummer: 313678-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...