Methyl-5-cyano-2-hydroxybenzenecarboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 5-cyano-2-hydroxybenzoate
OR31017-1G
OR31017-250MG
APOSOR31017-1GEA
82.8
EUR
APOSOR31017-1G
APOSOR31017-250MG
Methyl-5-cyano-2-hydroxybenzenecarboxylate
Methyl-5-cyano-2-hydroxybenzenecarboxylate
Formel:
C₉H₇NO₃ Molecular Weight: 177.16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00800949 CAS-Nummer: 84437-12-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...