5-Brom-1,4-benzodioxan 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2,3-dihydro-1,4-benzodioxane

OR310217-1G OR310217-5G
APOSOR310217-1GEA 127 EUR
APOSOR310217-1G APOSOR310217-5G
5-Brom-1,4-benzodioxan 96%
5-Brom-1,4-benzodioxan
Formel: C₈H₇BrO₂
Molecular Weight: 215,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095029
CAS-Nummer: 58328-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR