2-Chlorpyrimidin-5-carbonitril

Lieferant: Apollo Scientific
Achtung

OR310354-1G OR310354-25G OR310354-5G
APOSOR310354-1GEA 66.7 EUR
APOSOR310354-1G APOSOR310354-25G APOSOR310354-5G
2-Chlorpyrimidin-5-carbonitril
2-Chlorpyrimidin-5-carbonitril
Formel: C₅H₂ClN₃
Molecular Weight: 139,5400 g/mol
Schmelzpunkt: 140…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10697145
CAS-Nummer: 1753-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR