2-Vinylbenzoic acid
Lieferant: Apollo Scientific
|
Gefahr
|
OR31038-1G
OR31038-5G
OR31038-10G
APOSOR31038-1GEA
69.6
EUR
APOSOR31038-1G
APOSOR31038-5G
APOSOR31038-10G
2-Vinylbenzoic acid
2-Vinylbenzoic acid
Formel:
C₉H₈O₂ Molecular Weight: 148.16 g/mol Schmelzpunkt: 94 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02066273 CAS-Nummer: 27326-43-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...