tert-Butyl 2-phenylpiperazine-1-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Phenylpiperazine, N1-BOC protected, 1-Boc-2-phenylpiperazine, 1-(tert-Butoxycarbonyl)-2-phenylpiperazine, 2-Methyl-2-propanyl 2-phenyl-1-piperazinecarboxylate

OR3104-1G OR3104-250MG OR3104-100MG
APOSOR3104-1GEA 227 EUR
APOSOR3104-1G APOSOR3104-250MG APOSOR3104-100MG
tert-Butyl 2-phenylpiperazine-1-carboxylate
tert-Butyl 2-phenylpiperazine-1-carboxylate
Formel: C₁₅H₂₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07367734
CAS-Nummer: 886766-60-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR