N-Boc-3-ethylpiperazine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Ethylpiperazine, N1-BOC protected, N-Boc-3-Ethylpiperazine
OR3105-1G
OR3105-5G
APOSOR3105-1GEA
91.5
EUR
APOSOR3105-1G
APOSOR3105-5G
N-Boc-3-ethylpiperazine
N-Boc-3-ethylpiperazine
Formel:
C₁₁H₂₂N₂O₂ Molecular Weight: 214,3046 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07367735 CAS-Nummer: 438049-35-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...