5,5'-Dimethyl-2,2'-bipyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 5,5'-Dimethyl-2,2'-bipyridine, Abametapir, 6,6'-Bi-3-picoline

OR31053-5G OR31053-10G OR31053-25G
APOSOR31053-5GEA 26.1 EUR
APOSOR31053-5G APOSOR31053-10G APOSOR31053-25G
5,5'-Dimethyl-2,2'-bipyridin
5,5'-Dimethyl-2,2'-bipyridyl
Formel: C₁₂H₁₂N₂
Molecular Weight: 184,24 g/mol
Schmelzpunkt: 114…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01740554
CAS-Nummer: 1762-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR