(R)-1-Boc-2-methylpiperazin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2R)-2-Methylpiperazine, N1-BOC protected, (R)-tert-Butyl 2-methylpiperazine-1-carboxylate
OR3106-100G
OR3106-25G
OR3106-10G
APOSOR3106-100GEA
451
EUR
APOSOR3106-100G
APOSOR3106-25G
APOSOR3106-10G
(R)-1-Boc-2-methylpiperazin 98%
(R)-1-Boc-2-methylpiperazin
Formel:
C₁₀H₂₀N₂O₂ Molecular Weight: 200,2780 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862120 CAS-Nummer: 170033-47-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...