4-Cyanbenzophenon 97%
Lieferant: Apollo Scientific
Synonyme:
4-Benzoylbenzonitril
OR310642-1G
OR310642-5G
APOSOR310642-1GEA
81.2
EUR
APOSOR310642-1G
APOSOR310642-5G
4-Cyanbenzophenon 97%
4-Cyanbenzophenon
Formel:
C₁₄H₉NO Molecular Weight: 207.23 g/mol Siedepunkt: 363 Schmelzpunkt: 110…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016382 CAS-Nummer: 1503-49-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...