6-(4-Nitrophenyl)imidazo[2,1-b][1,3]thiazole

Lieferant: Apollo Scientific

OR310658-1G OR310658-5G
APOSOR310658-1GEA 270 EUR
APOSOR310658-1G APOSOR310658-5G
6-(4-Nitrophenyl)imidazo[2,1-b][1,3]thiazole
6-(4-Nitrophenyl)imidazo[2,1-b][1,3]thiazole
Formel: C₁₁H₇N₃O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00113357
CAS-Nummer: 7120-14-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR