5-Bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Amino-7-bromo-9-methyl-7-deazapurine

OR310814-1G OR310814-250MG
APOSOR310814-1GEA 145 EUR
APOSOR310814-1G APOSOR310814-250MG
5-Bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine
5-Bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine
Formel: C₇H₇BrN₄
Molecular Weight: 227,065 g/mol
Schmelzpunkt: 169…198 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD20486185
CAS-Nummer: 1337532-51-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR