4-Bromphenylboronsäureneopentylglycolester
Lieferant: Apollo Scientific
Synonyme:
2-(4-Bromophenyl)-5,5-dimethyl-1,3,2-dioxaborinane, 4-Bromobenzeneboronic acid neopentyl glycol ester
OR310880-5G
APOSOR310880-5GEA
92.8
EUR
APOSOR310880-5G
4-Bromphenylboronsäureneopentylglycolester
4-Bromphenylboronsäureneopentylglycolester
Formel:
C₁₁H₁₄BBrO₂ Molecular Weight: 268,94 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318105 CAS-Nummer: 183677-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...