7H-Pyrrolo[2,3-d]pyrimidine-2,4-diol

Lieferant: Apollo Scientific
Achtung

OR3109-1G OR3109-5G OR3109-25G
APOSOR3109-1GEA 29 EUR
APOSOR3109-1G APOSOR3109-5G APOSOR3109-25G
7H-Pyrrolo[2,3-d]pyrimidine-2,4-diol
7H-Pyrrolo[2,3-d]pyrimidine-2,4-diol
Formel: C₆H₅N₃O₂
Molecular Weight: 151.13 g/mol
Schmelzpunkt: >350 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03092515
CAS-Nummer: 39929-79-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR