5-Bromoindan-2-ylamine hydrobromide

Lieferant: Apollo Scientific
Achtung

OR310934-1G
APOSOR310934-1GEA 270 EUR
APOSOR310934-1G
5-Bromoindan-2-ylamine hydrobromide
5-Bromoindan-2-ylamine hydrobromide
Formel: C₉H₁₁Br₂N
Lagertemperatur: Kühlschrank
CAS-Nummer: 321352-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR