1,2,3,4-Tetrahydroisoquinolin-6-ol hydrobrimide

Lieferant: Apollo Scientific

OR310940-1G OR310940-500MG
APOSOR310940-1GEA 336 EUR
APOSOR310940-1G APOSOR310940-500MG
1,2,3,4-Tetrahydroisoquinolin-6-ol hydrobrimide
1,2,3,4-Tetrahydroisoquinolin-6-ol hydrobrimide
Formel: C₉H₁₂BrNO
Lagertemperatur: Raumtemperatur
CAS-Nummer: 59839-23-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR