3-Aminomethyl-3-[bis(phenylmethyl)amino]oxetane 97%

Lieferant: Apollo Scientific

OR312140-10G OR312140-1G OR312140-5G
APOSOR312140-10GEA 493 EUR
APOSOR312140-10G APOSOR312140-1G APOSOR312140-5G
3-Aminomethyl-3-[bis(phenylmethyl)amino]oxetane 97%
3-Aminomethyl-3-[bis(phenylmethyl)amino]oxetane
Formel: C₁₈H₂₂N₂O
Molecular Weight: 282,39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19443904
CAS-Nummer: 1021392-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR