trans-3-(Boc-amino)cyclobutanecarboxylic acid 98%
Lieferant: Apollo Scientific
Synonyme:
trans-3-(tert-butoxycarbonylamino)cyclobutanecarboxylic acid, (1r,3r)-3-((tert-Butoxycarbonyl)amino)cyclobutanecarboxylic acid, trans-3-((tert-Butoxycarbonyl)amino)cyclobutanecarboxylic acid
OR312151-1G
OR312151-5G
APOSOR312151-1GEA
115
EUR
APOSOR312151-1G
APOSOR312151-5G
trans-3-(Boc-amino)cyclobutanecarboxylic acid 98%
trans-3-(Boc-amino)cyclobutanecarboxylic acid
Formel:
C₁₀H₁₇NO₄ Molecular Weight: 215.249 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06657680 CAS-Nummer: 939400-34-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...