2,6-Diazaspiro[3.4]octane-2-carboxylic acid tert-butylester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 2,6-diazaspiro[3.4]octane-2-carboxylate, 2,6-Diazaspiro[3.4]octane, N2-BOC protected
OR312199-1G
OR312199-250MG
APOSOR312199-1GEA
226
EUR
APOSOR312199-1G
APOSOR312199-250MG
2,6-Diazaspiro[3.4]octane-2-carboxylic acid tert-butylester
2,6-Diazaspiro[3.4]octane-2-carboxylic acid tert-butylester
Formel:
C₁₁H₂₀N₂O₂ Molecular Weight: 212,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08234737 CAS-Nummer: 885270-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...